Tisk…

Anti-CD9 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26491-0.2EA 20890 CZK
BIRBORB26491-0.2
Anti-CD9 Mouse Monoclonal Antibody
Protilátky
  • Flow Cytometry: 2 μg; Immunochemistry: 1:25-1:50

Type: Primary
Antigen: CD9 (CD9 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR