Tisk…

Anti-ITGAM Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26501-0.2EA 0 CZK
BIRBORB26501-0.2
Anti-ITGAM Mouse Monoclonal Antibody
Protilátky
Flow Cytometry: 2 ?g /5x105; Immunochemistry: 1:25-1:50

Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR