Tisk…

Anti-CD19 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26507-0.2EA 0 CZK
BIRBORB26507-0.2
Anti-CD19 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR