Tisk…

Anti-GP1BA Mouse Monoclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB26531-100EA 0 CZK
BIRBORB26531-100
Anti-GP1BA Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Flow Cytometry: 20 ?l/5x105

Type: Primary
Antigen: GP1BA (glycoprotein Ib (platelet), alpha polypeptide)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR