Tisk…
Anti-TNFRSF8 Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB26533-0.2EA 0 CZK
BIRBORB26533-0.2
Anti-TNFRSF8 Mouse Monoclonal Antibody
Protilátky
Flow Cytometry: 2 ?g; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: TNFRSF8 (tumor necrosis factor receptor superfamily, member 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human