Tisk…

Anti-MAP-2 (2a + 2b) Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26626-0.25EA 0 CZK
BIRBORB26626-0.25
Anti-MAP-2 (2a + 2b) Mouse Monoclonal Antibody
Protilátky
Immunochemistry: 1:500

Type: Primary
Antigen: MAP-2 (2a + 2b)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR