Tisk…

Anti-SNAP25 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26633-1EA 0 CZK
BIRBORB26633-1
Anti-SNAP25 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SNAP25 (synaptosomal-associated protein, 25kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR