Tisk…

Anti-UB Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26656-100EA 0 CZK
BIRBORB26656-100
Anti-UB Mouse Monoclonal Antibody
Protilátky
ELISA: 1:10000-1:100000; Western Blot: 1:1000-1:100000; Immunoprecipitation: 1:10-1:100; Immunofluorescence: 1:100-1:10000

Type: Primary
Antigen: UB (ubiquitin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR