Tisk…

Anti-CAMP Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26668-20EA 0 CZK
BIRBORB26668-20
Anti-CAMP Rabbit Polyclonal Antibody
Protilátky
  • RIA: 1:1000

Type: Primary
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR