Tisk…

Anti-APP Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26677-0.2EA 0 CZK
BIRBORB26677-0.2
Anti-APP Rabbit Polyclonal Antibody
Protilátky
  • ELISA: 1:1000; Immunochemistry: 1:200

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR