Tisk…
Anti-Nicotinic alpha 7 Rec. Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB26707-100EA 0 CZK
BIRBORB26707-100
Anti-Nicotinic alpha 7 Rec. Rabbit Polyclonal Antibody
Protilátky
Immunochemistry: 1:400; Western Blot: 1:800
Type: Primary
Antigen: Nicotinic alpha 7 Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat