Tisk…

Anti-D4 Dopamine Rec. Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26719-100EA 0 CZK
BIRBORB26719-100
Anti-D4 Dopamine Rec. Rabbit Polyclonal Antibody
Protilátky
Immunochemistry: 1:10000; Western Blot: 1:500

Type: Primary
Antigen: D4 Dopamine Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR