Tisk…

Anti-Nicotinic beta 2 Rec. Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26720-100EA 0 CZK
BIRBORB26720-100
Anti-Nicotinic beta 2 Rec. Rabbit Polyclonal Antibody
Protilátky
Immunochemistry: 1:200; Western Blot: 1:400

Type: Primary
Antigen: Nicotinic beta 2 Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR