Tisk…
Anti-Endorphin, beta Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB26731-100EA 0 CZK
BIRBORB26731-100
Anti-Endorphin, beta Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Endorphin, beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat