Tisk…

Anti-NOS3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26742-100EA 0 CZK
BIRBORB26742-100
Anti-NOS3 Rabbit Polyclonal Antibody
Protilátky
Immunofluorescence: 1:400; Western Blot: 1:800

Type: Primary
Antigen: NOS3 (nitric oxide synthase 3 (endothelial cell))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR