Tisk…

Anti-Staph. Enterotoxin C Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26799-1EA 0 CZK
BIRBORB26799-1
Anti-Staph. Enterotoxin C Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Staph. Enterotoxin C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR