Tisk…

Anti-COL1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26855-0.5EA 0 CZK
BIRBORB26855-0.5
Anti-COL1 Rabbit Polyclonal Antibody
Protilátky
Immunofluorescence: 1:80; Immunochemistry: 1:500; ELISA: 1:200

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR