Tisk…

Anti-CA2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26890-50EA 0 CZK
BIRBORB26890-50
Anti-CA2 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:20000-1:115000

Type: Primary
Antigen: CA2 (carbonic anhydrase II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR