Tisk…

Anti-Elastase Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26892-50EA 0 CZK
BIRBORB26892-50
Anti-Elastase Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:1000-1:5400

Type: Primary
Antigen: Elastase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR