Tisk…

Anti-IgM Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB26914-5EA 0 CZK
BIRBORB26914-5
Anti-IgM Goat Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR