Tisk…

Anti-KRT7 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB27200-5EA 0 CZK
BIRBORB27200-5
Anti-KRT7 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR