Tisk…

Anti-MS4A1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB27261-1EA 0 CZK
BIRBORB27261-1
Anti-MS4A1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR