Tisk…

Anti-FLT4 Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB27269-50EA 0 CZK
BIRBORB27269-50
Anti-FLT4 Mouse monoclonal antibody
Protilátky
ELISA: 0.5-1.5 ?g/ml; Western Blot: 0.5-1 ?g/ml; Immunochemistry: not tested

Type: Primary
Antigen: FLT4 (fms-related tyrosine kinase 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR