Tisk…
Anti-AGE Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB27271-1EA 0 CZK
BIRBORB27271-1 BIRBORB27271-0.2
Anti-AGE Rabbit Polyclonal Antibody
Protilátky
ELISA: 1 ?g/ml; Immunochemistry: 5-10 ?g/ml
Type: Primary
Antigen: AGE
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat