Tisk…
Anti-AGE-1 Glucose modified Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB27285-10EA 0 CZK
BIRBORB27285-10
Anti-AGE-1 Glucose modified Mouse Monoclonal Antibody
Protilátky
WB/ ELISA: 0.1 ?g/ml
Type: Primary
Antigen: AGE-1 Glucose modified
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Objednat