Tisk…

Anti-Vitellogenin Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB27299-100EA 0 CZK
BIRBORB27299-100
Anti-Vitellogenin Mouse Monoclonal Antibody
Protilátky
Immunoblot: 0.1-1 ?g/ml; ELISA: 0.25-2 ?g/ml

Type: Primary
Antigen: Vitellogenin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Fish
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR