Tisk…

Anti-COL1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27310-0.5EA 0 CZK
BIRBORB27310-0.5 BIRBORB27310-0.1
Anti-COL1 Rabbit Polyclonal Antibody
Protilátky
ImmunofluorescenceA 1:80; Immunochemistry: 1:500; ELISA: >1:200

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR