Tisk…

Anti-Bombesin Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB27354-0.25EA 0 CZK
BIRBORB27354-0.25
Anti-Bombesin Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: Bombesin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR