Tisk…

Anti-Macrophages / Monocytes, MAC387 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB27425-5EA 0 CZK
BIRBORB27425-5
Anti-Macrophages / Monocytes, MAC387 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Macrophages / Monocytes, MAC387
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR