Tisk…
Anti-Mammaglobin A Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB27426-5EA 0 CZK
BIRBORB27426-5
Anti-Mammaglobin A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Mammaglobin A
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat