Tisk…

Anti-TP53 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB27454-5EA 0 CZK
BIRBORB27454-5
Anti-TP53 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR