Tisk…

Anti-TTF1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB27475-5EA 0 CZK
BIRBORB27475-5
Anti-TTF1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TTF1 (transcription termination factor, RNA polymerase I)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR