Tisk…

Anti-POMC Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27478-1EA 0 CZK
BIRBORB27478-1
Anti-POMC Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: POMC (proopiomelanocortin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR