Tisk…

Anti-Pyrralin Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB27505-20EA 0 CZK
BIRBORB27505-20
Anti-Pyrralin Mouse Monoclonal Antibody
Protilátky
ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: Pyrralin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR