Tisk…

Anti-AGER Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB27509-50EA 0 CZK
BIRBORB27509-50
Anti-AGER Mouse Monoclonal Antibody
Protilátky
Western Blot: 0.1 ?g/ml; Immunochemistry: 1 ?g/ml; FACS: 1 ?g/ml; ELISA: 0.01 ?g/ml

Type: Primary
Antigen: AGER (advanced glycosylation end product-specific receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR