Tisk…

Anti-IgG Horse Polyclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB27530-1EA 0 CZK
BIRBORB27530-1
Anti-IgG Horse Polyclonal Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: Mouse/Rabbit IgG
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Horse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR