Tisk…
Anti-ACTG1 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB27539-100EA 0 CZK
BIRBORB27539-100
Anti-ACTG1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ACTG1 (actin, gamma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Objednat