Tisk…

Anti-CAPN1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27577-100EA 0 CZK
BIRBORB27577-100
Anti-CAPN1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CAPN1 (calpain 1, (mu/I) large subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR