Tisk…

Anti-GJA5 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27581-100EA 0 CZK
BIRBORB27581-100
Anti-GJA5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GJA5 (gap junction protein, alpha 5, 40kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR