Tisk…

Anti-TIMP1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27597-100EA 0 CZK
BIRBORB27597-100
Anti-TIMP1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR