Tisk…
Anti-HSD17B6 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB27598-100EA 0 CZK
BIRBORB27598-100
Anti-HSD17B6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HSD17B6 (hydroxysteroid (17-beta) dehydrogenase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human