Tisk…

Anti-GAD2 None polyclonal antibody

Dodavatel: Biorbyt
BIRBORB27613-100EA 0 CZK
BIRBORB27613-100
Anti-GAD2 None polyclonal antibody
Protilátky
Type: Primary
Antigen: GAD2 (glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG (Rabbit)
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR