Tisk…

Anti-MAPK8/9 None Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27617-100EA 0 CZK
BIRBORB27617-100
Anti-MAPK8/9 None Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MAPK8/9 (mitogen-activated protein kinase 8/9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR