Tisk…

Anti-AHSG Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27630-100EA 0 CZK
BIRBORB27630-100
Anti-AHSG Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AHSG (alpha-2-HS-glycoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR