Tisk…

Anti-CA1 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB27634-100EA 0 CZK
BIRBORB27634-100
Anti-CA1 Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: CA1 (carbonic anhydrase I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR