Tisk…

Anti-CXCL9 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27638-100EA 0 CZK
BIRBORB27638-100
Anti-CXCL9 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CXCL9 (chemokine (C-X-C motif) ligand 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR