Tisk…

Anti-CASP6 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB27642-100EA 0 CZK
BIRBORB27642-100
Anti-CASP6 Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: CASP6 (caspase 6, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR