Tisk…

Anti-FGF22 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27673-100EA 0 CZK
BIRBORB27673-100
Anti-FGF22 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FGF22 (fibroblast growth factor 22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR