Tisk…

Anti-IgG Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27710-1EA 0 CZK
BIRBORB27710-1 BIRBORB27710-0.5
Anti-IgG Goat Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR