Tisk…

Anti-Protein A None Antibody

Dodavatel: Biorbyt
BIRBORB27715-1EA 0 CZK
BIRBORB27715-1 BIRBORB27715-0.5
Anti-Protein A None Antibody
Protilátky
Type: Secondary
Antigen: Protein A
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR